Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518754
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518754 Supplier Invitrogen™ Supplier No. PA518754
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDE
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene IDE
Gene Accession No. P35559, Q9JHR7
Gene Alias 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603
Gene Symbols IDE
Host Species Goat
Immunogen Synthetic peptide sequence (QQYNYDRDNIE) corresponding to the internal amino acids of Ide (aa 914-924).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15925, 25700
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.