Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH1 (R132H) Recombinant Rabbit Monoclonal Antibody (RM390)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544585
Description
IDH1 (R132H) Recombinant Monoclonal Antibody for IHC (P)
Isocitrate dehydrogenases such as IDH1 catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the cytoplasm and peroxisomes. It contains the PTS-1 peroxisomal targeting signal sequence. The presence of this enzyme in peroxisomes suggests roles in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as in peroxisomal reactions that consume 2-oxoglutarate, namely the alpha-hydroxylation of phytanic acid. The cytoplasmic enzyme serves a significant role in cytoplasmic NADPH production. This product is specific for IDH1 (mutant R132H).
Specifications
IDH1 (R132H) | |
Recombinant Monoclonal | |
100 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
O75874 | |
IDH1 | |
A peptide corresponding to human (R132H mutant specific). The immunogen range is a synthetic peptide within amino acids 180-280 of human IDH1 with R132H mutant. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
RM390 | |
Unconjugated | |
IDH1 | |
Cytosolic NADP-isocitrate dehydrogenase; epididymis luminal protein 216; epididymis secretory protein Li 26; HEL-216; HEL-S-26; IDCD; IDH; IDH1; IDH1-mutant; IDP; IDPC; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; isocitrate dehydrogenase [NADP] cytoplasmic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; NADP(+)-specific ICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; oxalosuccinate decarboxylase; PICD | |
Rabbit | |
Protein A | |
RUO | |
3417 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction