Learn More
Invitrogen™ IDH2 Monoclonal Antibody (6B13)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence identical to the related mouse and rat sequences. Positive Control - WB: human Hela whole cell, human Sw620 whole cell, human MCF-7 whole cell, human HEPG2 whole cell, human Jurkat whole cell, rat heart tissue, rat liver tissue, rat PC-12 whole cell, mouse heart tissue, mouse liver tissue, mouse NIH/3T3 whole cell. IHC: human testicular cancer tissue, human rectal cancer tissue, human melanoma tissue, human ovarian cancer tissue, rat skeletal muscle tissue. ICC/IF: A431 cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | IDH2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6B13 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | Idh2 |
| Gene Accession No. | P48735, P54071, P56574 |
| Gene Alias | D2HGA2; E430004F23; ICD-M; IDH; Idh2; Idh-2; IDHM; IDP; IDPM; isocitrate dehydrogenase (NADP(+)) 2; isocitrate dehydrogenase (NADP(+)) 2, mitochondrial; isocitrate dehydrogenase [NADP], mitochondrial; isocitrate dehydrogenase 2 (NADP+), mitochondrial; mNADP-IDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP+-specific isocitrate dehydrogenase; NADPH-specific isocitrate dehydrogenase; oxalosuccinate decarboxylase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.