Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDH2 Mouse anti-Human, Unconjugated, Clone: 5F11, Abnova™

Catalog No. 89335104 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-104 Supplier Abnova Corporation Supplier No. H00003418M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

IDH2 Monoclonal antibody specifically detects IDH2 in Human samples. It is validated for IHC-P, S-ELISA, ELISA, WB-Re, WB-Tr.

Specifications

Antigen IDH2
Applications Immunohistochemistry (Paraffin), ELISA, Western Blot
Classification Monoclonal
Clone 5F11
Conjugate Unconjugated
Description isocitrate dehydrogenase 2 (NADP+), mitochondrial
Gene IDH2
Gene Accession No. NM_002168
Gene Alias ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
Host Species Mouse
Immunogen IDH2 (NP_002159, 354 a.a. ∼ 451 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3418
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.