Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596605 Supplier Invitrogen™ Supplier No. PA596605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KMVFTPKDGS GVKEWEVYNF PAGGVGMGMY NTDESISGFA HSCFQYAIQK KWPLYMSTKN TILKAYDGRF KDIFQEIFDK HYKTDFDKNK IWYEHRLIDD MVAQVLKSSG GFVWACKNYD GDVQSDILAQ GFGSLGLMTS VLVCPDGKTI EAEAAHGTVT RHYREHQKGR PTSTNPIASI FAWTRGLEHR GKLDGNQDLI RFAQMLEKVC VETVESGAMT KDLAGCIHGL SNVKLNEHFL NTTDFLDTIK SNLDRALGRQ; Positive Samples: HL-60, MCF-7, Jurkat, HepG2, U-87MG, Mouse kidney, Mouse heart, Rat lung, Rat kidney; Cellular Location: Mitochondrion.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NADas the electron acceptor and the other NADP. Five isocitrate dehydrogenases have been reported: three NAD-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH2
Applications ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Idh2
Gene Accession No. P48735, P54071, P56574
Gene Alias D2HGA2; E430004F23; ICD-M; IDH; Idh2; Idh-2; IDHM; IDP; IDPM; isocitrate dehydrogenase (NADP(+)) 2; isocitrate dehydrogenase (NADP(+)) 2, mitochondrial; isocitrate dehydrogenase [NADP], mitochondrial; isocitrate dehydrogenase 2 (NADP+), mitochondrial; mNADP-IDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP+-specific isocitrate dehydrogenase; NADPH-specific isocitrate dehydrogenase; oxalosuccinate decarboxylase
Gene Symbols Idh2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 193-452 of human IDH2 (NP_0021592).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269951, 3418, 361596
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.