Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519412
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NADas the electron acceptor and the other NADP. Five isocitrate dehydrogenases have been reported: three NAD-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex.
Specifications
| IDH2 | |
| Polyclonal | |
| Unconjugated | |
| Idh2 | |
| D2HGA2; E430004F23; ICD-M; IDH; Idh2; Idh-2; IDHM; IDP; IDPM; isocitrate dehydrogenase (NADP(+)) 2; isocitrate dehydrogenase (NADP(+)) 2, mitochondrial; isocitrate dehydrogenase [NADP], mitochondrial; isocitrate dehydrogenase 2 (NADP+), mitochondrial; mNADP-IDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP+-specific isocitrate dehydrogenase; NADPH-specific isocitrate dehydrogenase; oxalosuccinate decarboxylase | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 269951, 3418, 361596, 397603 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P33198, P48735, P54071, P56574 | |
| Idh2 | |
| Synthetic peptide sequence (CIHGLSNVKLNE) corresponding to the internal amino acids of IDH2 (aa 418-429). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction