Learn More
IDH2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304096AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells. For IHC, epitope retrieval with c
Plays a role in intermediary metabolism and energy production. It may tightly associate or interact with the pyruvate dehydrogenase complex.Specifications
IDH2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P48735, P54071 | |
Idh2 | |
Between 402 and 452 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
IDH2 | |
IDH2; isocitrate dehydrogenase 2 (NADP+), mitochondrial; ICD-M; IDH; IDHM; IDP; IDPM; mNADP-IDH; NADP+-specific ICDH; isocitrate dehydrogenase, mitochondrial; oxalosuccinate decarboxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
269951, 3418 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.