Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH3A Monoclonal Antibody (OTI2F11)
GREENER_CHOICE

Catalog No. PIMA525007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525007 Supplier Invitrogen™ Supplier No. MA525007
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IDH3A Monoclonal Antibody for Western Blot, ICC/IF, Flow

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein described here is the alpha subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH3A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IDH3A
Gene Accession No. P50213, P54836
Gene Alias 1110003P10Rik; 1500012E04Rik; AA407078; AI316514; H-IDH alpha; Idh3a; isocitrate dehydrogenase (NAD(+)) 3 alpha; isocitrate dehydrogenase (NAD(+)) 3 catalytic subunit alpha; isocitrate dehydrogenase (NAD+) alpha chain; isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) alpha; isocitrate dehydrogenase 3 (NAD+) alpha; isocitric dehydrogenase subunit alpha; NAD(+)-specific ICDH subunit alpha; NAD(H)-specific isocitrate dehydrogenase alpha subunit; NAD+-specific ICDH
Gene Symbols IDH3A
Host Species Mouse
Immunogen Full length human recombinant protein of IDH3A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3419, 479066
Target Species Human, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.