Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583559
Description
Immunogen sequence: PTALLLSAVM MLRHMGLFDH AARIEAACFA TIKDGKSLTK DLGGNAKCSD FTEEICRRV.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein described here is the alpha subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase.
Specifications
IDH3A | |
Polyclonal | |
Unconjugated | |
IDH3A | |
1110003P10Rik; 1500012E04Rik; AA407078; AI316514; H-IDH alpha; Idh3a; isocitrate dehydrogenase (NAD(+)) 3 alpha; isocitrate dehydrogenase (NAD(+)) 3 catalytic subunit alpha; isocitrate dehydrogenase (NAD+) alpha chain; isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) alpha; isocitrate dehydrogenase 3 (NAD+) alpha; isocitric dehydrogenase subunit alpha; NAD(+)-specific ICDH subunit alpha; NAD(H)-specific isocitrate dehydrogenase alpha subunit; NAD+-specific ICDH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3419 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50213 | |
IDH3A | |
Recombinant protein corresponding to Human IDH3A. Recombinant protein control fragment (Product #RP-98112). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction