Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550337
Description
The antibody detects endogenous levels of total IDH3B protein.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
IDH3B | |
Polyclonal | |
Unconjugated | |
Idh3b | |
C78231; DH3B; H-IDHB; H-IDHG; IDH3B; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) beta; isocitrate dehydrogenase 3 (NAD+) beta; isocitrate dehydrogenase 3 beta; isocitric dehydrogenase subunit beta; N14A tumor-related protein; NAD(+)-specific ICDH subunit beta; NAD+-specific isocitrate dehydrogenase b subunit; RP46 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170718, 3420 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O43837 | |
Idh3b | |
Fusion protein corresponding to residues near the C terminal of human isocitrate dehydrogenase 3 (NAD+) gamma. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction