Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDH3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575756
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
IDH3B | |
Polyclonal | |
Unconjugated | |
Idh3b | |
C78231; DH3B; H-IDHB; H-IDHG; IDH3B; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) beta; isocitrate dehydrogenase 3 (NAD+) beta; isocitrate dehydrogenase 3 beta; isocitric dehydrogenase subunit beta; N14A tumor-related protein; NAD(+)-specific ICDH subunit beta; NAD+-specific isocitrate dehydrogenase b subunit; RP46 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170718, 3420, 94173 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43837, Q68FX0 | |
Idh3b | |
Synthetic peptide corresponding to residues in Human IDH3B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction