Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589402 Supplier Invitrogen™ Supplier No. PA589402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASRSQAEDVR VEGSFPVTML PGDGVGPELM HAVKEVFKAA AVPVEFQEHH LSEVQNMASE EKLEQVLSSM KENKVAIIGK IHTPMEYKGE LASYDMRLRR KLDLFANVVH VKSLPGYMTR HNNLDLVIIR EQTEGE; Positive Samples: H460, HeLa, 293T, Mouse kidney, Mouse heart, Mouse lung, Rat kidney, Rat heart, Rat lung; Cellular Location: Mitochondrion.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH3B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Idh3b
Gene Accession No. O43837, Q68FX0
Gene Alias C78231; DH3B; H-IDHB; H-IDHG; IDH3B; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) beta; isocitrate dehydrogenase 3 (NAD+) beta; isocitrate dehydrogenase 3 beta; isocitric dehydrogenase subunit beta; N14A tumor-related protein; NAD(+)-specific ICDH subunit beta; NAD+-specific isocitrate dehydrogenase b subunit; RP46
Gene Symbols Idh3b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-170 of human IDH3B (NP_0088302).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170718, 3420, 94173
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.