Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDH3G Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$416.50 - $682.00
Specifications
Antigen | IDH3G |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IDH3G Polyclonal specifically detects IDH3G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IDH3G | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
EC 1.1.1, EC 1.1.1.41, H-IDHG, IDH-gamma, isocitrate dehydrogenase 3 (NAD+) gamma, isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit, mitochondrial, NAD (H)-specific isocitrate dehydrogenase gamma subunit, NAD+-specific ICDH | |
IDH3G | |
IgG | |
Affinity Purified | |
Specificity of human IDH3G antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3421 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ADEEDIRNAIMAIRRNRVALKGNIETNHNLPPSHKSRNNILRTSLDLYANVIHCKSLPGVVTRHKDIDILIVRENTEGEYSSLEHESVAGVVESLKIITKAKSLRIAEYAFKLAQESGRKKVTAVHKANIMKLGDGLFLQCCREVAA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title