Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH3G Monoclonal Antibody (2A2-1D3)
GREENER_CHOICE

Catalog No. PIMA519176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519176 Supplier Invitrogen™ Supplier No. MA519176
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MALKVATVAG SAAKAVLGPA LLCRPWEVLG AHEVPSRNIF SEQTIPPSAK YGGRHTVTMI PGDGIGPELM LHVKSVFRHA CVPVDFEEVH VSSNADEEDI RNAIMAIRRN RVALKGNIET NHNLPPSHKS RNNILRTSLD LYANVIHCKS LPGVVTRHKD IDILIVRENT EGEYSSLEHE SVAGVVESLK IITKAKSLRI AEYAFKLAQE SGRKKVTAVH KANIMKLGDG LFLQCCREVA ARYPQITFEN MIVDNTTMQL VSRPQQFDVM VMPNLYGNIV NNVCAGLVGG PGLVAGANYG HVYAVFETAT RNTGKSIANK NIANPTATLL ASCMMLDHLK LHSYAASIRK AVLASMDNEN MHTPDIGGQG TTSEAIQDVI RHIRVINGRA VEA.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia. Several alternatively spliced transcript variants of this gene have been described, but only some of their full length natures have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH3G
Applications ELISA
Classification Monoclonal
Clone 2A2-1D3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Idh3g
Gene Accession No. P51553
Gene Alias H-IDHG; IDH; IDH3G; IDH-gamma; iso; Isocitrate dehydrogenase [NAD] subunit gamma 1, mitochondrial; isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) gamma; isocitrate dehydrogenase 3 (NAD), gamma; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 (NAD+), gamma; isocitrate dehydrogenase 3 gamma; isocitrate dehydrogenase 3, gamma; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-isocitrate dehydrogenase gamma subunit; NAD+-specific ICDH
Gene Symbols Idh3g
Host Species Mouse
Immunogen IDH3G (AAH00933, 1 a.a. ~ 393 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3421
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.