Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580704
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IDI2/IPPI2.
Catalyzes the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP).
Specifications
IDI2 | |
Polyclonal | |
Unconjugated | |
IDI2 | |
4833405L16Rik; diphosphate dimethylallyl diphosphate isomerase 2; Gm9745; Idi2; IPP isomerase 2; IPPI2; isopentenyl diphosphate delta-isomerase type 2; isopentenyl pyrophosphate isomerase 2; isopentenyl-diphosphate delta isomerase 2; isopentenyl-diphosphate Delta-isomerase 2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
91734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9BXS1 | |
IDI2 | |
Recombinant Human IDI2/IPPI2 Protein (Met1-Val227). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction