Learn More
IDN3Goat anti-Human, Polyclonal, Rockland™

Goat Polyclonal Antibody
Supplier: Rockland Immunochemicals 600101288

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human IDN3 protein. A BLAST analysis was used to suggest reactivity with this protein from human, rat, mouse, dog, bovine and orangutan based on 100% homology for the immunogen sequence. Expect reactivity with multiple splice variants of this protein. Cross reactivity with IDN3 homologues from other sources has not been determined.
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.Specifications
IDN3 | |
Polyclonal | |
Unconjugated | |
NIPBL | |
4921518A06Rik; 4933421G18Rik; C79399; CDLS; CDLS1; Delangin; Delangin homolog; ID; IDN3; IDN3-B; Nipbl; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); Nipped-B-like protein; Scc2; SCC2 homolog; sister chromatid cohesion 2 homolog | |
Goat | |
RUO | |
25836 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1.6 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Q6KC79 | |
NIPBL | |
100 μg | |
Primary | |
Human | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.