Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDNK Monoclonal Antibody (1H4)
GREENER_CHOICE

Catalog No. PIMA523317
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523317 Supplier Invitrogen™ Supplier No. MA523317
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KKTYRDILTQ GKDGVALKCE ESGKEAKQAE MQLLVVHLSG SFEVISGRLL KREGHFMPPE LLQSQFETLE PPAAPENFIQ ISVDKNVSEI IATIMETLKM K.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDNK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Idnk
Gene Accession No. Q5T6J7
Gene Alias 5133401N09Rik; bA522I20.2; BCRG2; C9orf103; glucokinase-like protein; gluconate kinase; gluconokinase-like protein; hGntK; Idnk; idnK gluconokinase homolog (E. coli); IDNK, gluconokinase; idnK, gluconokinase homolog; KIAA1824; probable gluconokinase; WDR22
Gene Symbols Idnk
Host Species Mouse
Immunogen IDNK (NP_001001551, 41 a.a. ~ 141 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414328
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.