Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO-2 Mouse anti-Mouse, MilliporeSigma™

Catalog No. MABF205525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2055-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2055-25 Supplier MilliporeSigma Supplier No. MABF205525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IDO-2, clone 4-3 is a mouse monoclonal antibody that detects IDO2 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDO-2
Applications Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Ido2,Indol1
Host Species Mouse
Immunogen KLH-conjugated linear peptide/recombinant protein corresponding to 21 amino acids from the C-terminal region of murine Indoleamine 2,3-dioxygenase 2 (IDO-2).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 4-3 detects IDO-2 in mouse. It targets an epitope within 21 amino acids in the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.