Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO1 Monoclonal Antibody (10.1), Rockland™
SDP

Catalog No. 501990586
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0586 100 μg
50-199-0587 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0586 Supplier Rockland Immunochemicals Supplier No. 200301E57
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human and mouse IDO1 protein. A BLAST analysis was used to suggest cross-reactivity with IDO1 from human and mouse sources based on 100% homology with the immunizing sequence. Cross-reactivity with IDO1 from other sources has not been determined.

IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.

Specifications

Antigen IDO1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10.1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Ido1
Gene Accession No. P14902, P28776
Gene Alias EC 1.13.11.52; IDO; Ido1; IDO-1; Indo; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; Indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; Indoleamine-pyrrole 2,3-dioxygenase
Gene Symbols Ido1
Host Species Mouse
Immunogen Anti-IDO1 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with fragment of recombinant human and mouse IDO1 protein followed by hybridoma development.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15930, 3620
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.