Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDO1 Mouse anti-Human, Alexa Fluor 488, Clone: V50-1886, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567854
Description
IDO1 Monoclonal antibody specifically detects IDO1 in Human samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
| IDO1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| IDO; IDO-1; INDO; indoleamine 2,3-dioxygenase 1 | |
| Human IDO1 Recombinant Protein | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry | |
| V50-1886 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction