Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO1 Mouse anti-Human , Alexa Fluor 647, Clone: V50-1886, BD Biosciences™

Catalog No. BDB566648 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566648 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566648 Supplier BD Biosciences Supplier No. 566648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IDO1 Monoclonal antibody specifically detects IDO1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDO1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone V50-1886
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IDO; IDO-1; INDO; indoleamine 2,3-dioxygenase 1
Host Species Mouse
Immunogen Human IDO1 Recombinant Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.