Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDO1 Mouse anti-Human, BV480, Clone: V50-1886, BD Horizon™
Mouse Monoclonal Antibody
$268.00 - $472.00
Specifications
| Antigen | IDO1 |
|---|---|
| Clone | V50-1886 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 480 |
Description
IDO1 Monoclonal antibody specifically detects IDO1 in Human samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)Specifications
| IDO1 | |
| Flow Cytometry | |
| Brilliant Violet 480 | |
| Mouse | |
| Human | |
| IDO; IDO-1; INDO; indoleamine 2,3-dioxygenase 1 | |
| IgG1 κ | |
| Affinity Chromatography |
| V50-1886 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IDO1 Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title