Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDO1, Mouse anti-Human, Clone: ID 177, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16027
Specifications
IDO1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4. | |
CD107B/IDO/INDO | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
ID 177 | |
ELISA (1:2000-10000) Western Blot (1:2000-5000) The optimal working dilution should be determined by the end user. | |
indoleamine 2,3-dioxygenase 1 | |
IDO1 | |
Recombinant protein corresponding to human IDO1. | |
RUO | |
3620 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction