Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDO1 Mouse anti-Human, Mouse, Rat, Clone: 3G2G11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IDO1 |
---|---|
Clone | 3G2G11 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6949
![]() ![]() |
Proteintech Group Inc
665281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregcy, autoimmunity, transplantation, and neoplasia.Specifications
IDO1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD107B, IDO, IDO 1, IDO1, INDO, indoleamine 2,3 dioxygenase 1 | |
Ido1 | |
IgG1 | |
Protein G | |
Antibody |
3G2G11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P14902, P28776, Q9ERD9 | |
15930, 3620, 66029 | |
IDO1 Fusion Protein Ag3953 | |
Primary | |
-20°C | |
IDO1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title