Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IDO2 Antibody (OTI1A4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272453PE
Description
IDO2 Monoclonal specifically detects IDO2 in Human samples. It is validated for Flow Cytometry.Specifications
IDO2 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
IDO2 | |
Full length recombinant protein of human IDO2 (NP_919270) produced in HEK293T cells. | |
0.1 mL | |
169355.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1A4 | |
Flow Cytometry | |
EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction