Learn More
IDS Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI4G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504277

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Specifications
IDS | |
Monoclonal | |
Unconjugated | |
IDS | |
MPS2, SIDS | |
Mouse | |
Affinity Chromatography | |
RUO | |
15931, 3423, 363513, 492194 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P22304, Q08890 | |
IDS | |
Full length human recombit protein of human IDS produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.