Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDUA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545483 Supplier Invitrogen™ Supplier No. PA545483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DPVAAAPRPL PAGGRLTLRP ALRLPSLLLV HVCARPEKPP GQVTRLRALP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Rat: 80%.

IDUA encodes an enzyme that hydrolyzes the terminal alpha-L-iduronic acid residues of two glycosaminoglycans, dermatan sulfate and heparan sulfate. This hydrolysis is required for the lysosomal degradation of these glycosaminoglycans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDUA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IDUA
Gene Accession No. P35475
Gene Alias 6030426D08; Alpha-L-iduronidase; IDA; IDUA; iduronidase alpha-L-; iduronidase, alpha-L-; MPS1
Gene Symbols IDUA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human IDUA (aa 509-558).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3425
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.