Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iduronate 2-Sulfatase/IDS Antibody (OTI1D9), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270994JF549
Description
Iduronate 2-Sulfatase/IDS Monoclonal specifically detects Iduronate 2-Sulfatase/IDS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Iduronate 2-Sulfatase/IDS | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IDS | |
Full length recombinant protein of human Iduronate 2-Sulfatase/IDS (NP_000193) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
3423.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction