Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Iduronate 2 Sulfatase Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA546990
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse IDS is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.
Specifications
Iduronate 2 Sulfatase | |
Polyclonal | |
Unconjugated | |
Ids | |
alpha-L-iduronate sulfate sulfatase; AW214631; Ids; Iduronate 2-sulfatase; iduronate 2-sulfatase (Hunter syndrome); iduronate 2-sulfatase 14 kDa chain; Iduronate 2-sulfatase 42 kDa chain; iduronate sulfatase; iduronate-2-sulfatase; idursulfase; MPS2; SIDS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3423 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P22304 | |
Ids | |
Mouse myeloma cell line NS0-derived recombinant human IDS Ser26-Pro550. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction