Learn More
Cytiva IEF/SDS Sample Application Strips
Catalog No. 45001795 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5
Catalog No. Quantity
45-001-795 5
1 options
Catalog No. 45-001-795 Supplier Cytiva Supplier No. 18100226
Specifications
For Use With (Application) IEF/SDS sample
Type IEF/SDS Sample Application Strip (52 samples, 1-20 μl)
Quantity 5