Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IEICO-4F, Thermo Scientific Chemicals

Catalog No. H66546MD
Click to view available options
Quantity:
100 mg
250 mg
500 mg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2089044-02-8
Molecular Formula C103H100F4N4O4S4
Molecular Weight (g/mol) 1662.19
InChI Key JNJCNYUQCFAGFR-UHFFFAOYNA-N
Synonym 2,2 '-((2 Z,2 'Z)-(((4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-sindaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene-5,2- diyl))bis(methanylylidene))bis(5,6-difluoro-3-oxo-2,3-dihydro-1 H-indene-2,1- diylidene))dimalononitrile
IUPAC Name 2-[2-({5-[15-(5-{[1-(dicyanomethylidene)-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-2-ylidene]methyl}-3-[(2-ethylhexyl)oxy]thiophen-2-yl)-9,9,18-tris(4-hexylphenyl)-18-methyl-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl]-4-[(2-ethylhexyl)oxy]thiophen-2-yl}methylidene)-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene]propanedinitrile
SMILES CCCCCCC1=CC=C(C=C1)C1(C)C2=C(SC(=C2)C2=C(OCC(CC)CCCC)C=C(S2)C=C2C(=O)C3=CC(F)=C(F)C=C3C2=C(C#N)C#N)C2=C1C=C1C3=C(C=C(S3)C3=C(OCC(CC)CCCC)C=C(S3)C=C3C(=O)C4=CC(F)=C(F)C=C4C3=C(C#N)C#N)C(C1=C2)(C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1

Specifications

Quantity 250 mg
Formula Weight 1808.40
Physical Form Solid
Chemical Name or Material IEICO-4 F
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.