Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IEM 1925 dihydrobromide
SDP

Catalog No. 419850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-985-0 50 mg
41-981-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-985-0 Supplier Tocris Bioscience™ Supplier No. 4198/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Voltage- and use-dependent open-channel antagonist of AMPA receptors

IEM 1925 dihydrobromide is a voltage- and use-dependent open-channel antagonist of AMPA receptors. Selective between subtypes; blocks GluA2 subunit-lacking receptors more potently than GluA2-containing receptors (KD for GluA2-containing AMPAR is 210 times higher at -80 mV). More potent than IEM 1460 (Cat. No. 1636) and IEM 1754 due to a slower unblocking rate. Alleviates inflammatory pain in a rat model of peripheral inflammation.

Chemical Identifiers

CAS 258282-23-4
Molecular Formula C17H30Br2N2

Specifications

Quantity 50 mg
Formula Weight 422.24
Purity 95%
Molecular Weight (g/mol) 422.249
Chemical Name or Material IEM 1925 dihydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.