Learn More
Invitrogen™ IER2 Monoclonal Antibody (3E2)

Description
Peptide Sequence: MEVQKEAQRI MTLSVWKMYH SRMQRGGLRL HRSLQLSLVM RSARELYLSA KVEALEPEVS LPAALPSDPR LHPPREAEST AETATPDGEH PFPEPMDTQE APTAEETSAC CAPRPAKVSR KRRSSSLSDG GDAGLVPSKK ARLEEKEEEE GASSEVADRL QPPPAQAEGA FPNLARVLQR RFSGLLNCSP AAPPTAPPAC EAKPACRPAD SMLNVLVRAV VAF.
Specifications
Specifications
| Antigen | IER2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | IER2 |
| Gene Accession No. | Q9BTL4 |
| Gene Alias | AI317238; Ch1; CHX1; chxI protein homolog; cycloheximide-induced gene protein; ETR101; Growth factor-inducible immediate early protein; Ier2; immediate early response 2; immediate early response gene 2 protein; PIP92; proline-rich-induced protein 92; Protein ETR101; T-lymphocyte-activated protein |
| Gene Symbols | IER2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.