Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565948 Supplier Invitrogen™ Supplier No. PA565948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEVQKEAQRI MTLSVWKMYH SRMQRGGLRL HRSLQ.

DNA-binding protein that seems to act as a transcription factor (PubMed:19584537). Involved in the regulation of neuronal differentiation, acts upon JNK-signaling pathway activation and plays a role in neurite outgrowth in hippocampal cells. May mediate with FIBP FGF-signaling in the establishment of laterality in the embryo. Promotes cell motility, seems to stimulate tumor metastasis (PubMed:22120713). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IER2
Gene Accession No. Q9BTL4
Gene Alias AI317238; Ch1; CHX1; chxI protein homolog; cycloheximide-induced gene protein; ETR101; Growth factor-inducible immediate early protein; Ier2; immediate early response 2; immediate early response gene 2 protein; PIP92; proline-rich-induced protein 92; Protein ETR101; T-lymphocyte-activated protein
Gene Symbols IER2
Host Species Rabbit
Immunogen Recombinant Human IER2. Recombinant protein control fragment (Product #RP-106630).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.