Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IER3IP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569453
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a small protein that is localized to the endoplasmic reticulum (ER) and may play a role in the ER stress response by mediating cell differentiation and apoptosis. Transcription of this gene is regulated by tumor necrosis factor alpha and specificity protein 1 (Sp1). Mutations in this gene may play a role in microcephaly, epilepsy, and diabetes syndrome (MEDS), and a pseudogene of this gene is located on the long arm of chromosome 12.
Specifications
IER3IP1 | |
Polyclonal | |
Unconjugated | |
IER3IP1 | |
1110057H19Rik; 796; AI644142; AL022842; AL022863; AV026606; HSPC039; IER3IP1; immediate early response 3 interacting protein 1; immediate early response 3-interacting protein 1; MEDS; PRO2309; uncharacterized protein LOC500270 | |
Rabbit | |
Affinity chromatography | |
RUO | |
66191 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CR20 | |
IER3IP1 | |
synthetic peptide corresponding to a region of mouse IER3IP1 (aa 9-58). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction