Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IER5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120642 Supplier Invitrogen™ Supplier No. PA5120642
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, A-549, Mouse testis, Rat lung. Immunogen sequence: APQPAEDEPP APPAVCPRKR CAAGVGGGPA GCPAPGSTPL KKPRRNLEQP PSGGEDDDAE EMETGNVANL ISIFGSSFSG LLRKSPGGGR EEEEGEESGP E.

This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IER5
Gene Accession No. O89113, Q5VY09
Gene Alias Ier5; immediate early response 5; immediate early response gene 5 protein; immediately early response; MGC102760; PP4583; SBBI48; si:dz129i22.1; Unknown (protein for MGC:136422); wu:fa99a02; wu:fb04b03; wu:fb11f08; zgc:136422
Gene Symbols IER5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human IER5 (NP_057629.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15939, 498256, 51278
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.