Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IER5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA556287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556287 Supplier Thermo Scientific Supplier No. PA556287

Rabbit Polyclonal Antibody

Immunogen sequence: RRNLEQPPSG GEDDDAEEME TGNVANLISI FGSSFSGLLR KSPGGGREEE EGEESGPEAA EPGQICCDKP V Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 77%.

This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER5
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IER5
Gene Accession No. Q5VY09
Gene Alias Ier5; immediate early response 5; immediate early response gene 5 protein; immediately early response; MGC102760; PP4583; SBBI48; si:dz129i22.1; Unknown (protein for MGC:136422); wu:fa99a02; wu:fb04b03; wu:fb11f08; zgc:136422
Gene Symbols IER5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IER5
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51278
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.