Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IER5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570176 Supplier Invitrogen™ Supplier No. PA570176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

IER5L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER5L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IER5L
Gene Accession No. Q99J55
Gene Alias 2610524G09Rik; bA247A12.2; Ier5l; immediate early response 5 like; immediate early response 5-like; immediate early response gene 5-like protein; LOW QUALITY PROTEIN: immediate early response gene 5-like protein
Gene Symbols IER5L
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse IER5L (aa 331-380).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72500
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.