Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IEX-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548274
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548274 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548274 Supplier Invitrogen™ Supplier No. PA548274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IEX-1 Polyclonal Antibody for Western Blot

IEX-1 is a stress inducible gene that is induced by ionizing radiation, ultraviolet radiation, and a variety of growth factors, i.e., FAS and TNF-a. IEX-1 is widely expressed in epithelial and endocrine tissues, as well as in vascular endothelium. It plays an important role in the regulation of cellular growth, cell death and oncogenesis. IEX-1 is precisely regulated by multiple transcription factors such as p53, NF-kappa-B/rel, Sp1 and c-Myc, to ensure rapid and transient expression of IEX-1 in cells under a variety of stress conditions. IEX-1 is expressed as both a longer form (IEX1L) and a splice variant, designated IEX1S. It is localized to the nucleus and perinuclear region. Overexpression of IEX-1 facilitates apoptosis and cell cycle progression, whereas disruption of IEX-1 expression is associated with decreases in both apoptosis and cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IEX-1
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene IER3
Gene Accession No. P46694, P46695
Gene Alias AI663993; anti-death protein; cAMP inducible gene 3; cI-3; DADB-118P11.5; Dif2; DIF-2; differentiation-dependent gene 2 protein; expressed in pancreatic carcinoma; GLY96; gly96, mouse, homolog of; IER3; IEX1; IEX-1; IEX-1L; immediate early protein GLY96; immediate early response 3; immediate early response 3 protein; immediately early gene X-1; Ler3; PACAP-responsive gene 1 protein; PRG1; Protein DIF-2; Protein PRG1; Radiation-inducible immediate-early gene IEX-1
Gene Symbols IER3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 38-65 amino acids from the N-terminal region of human IER3.
Purification Method Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15937, 8870
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.