Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ifenprodil Hemitartrate, 99%, PROMO, Thermo Scientific Chemicals

Catalog No. J60932MFPM
Click to view available options
Quantity:
50 mg

NMDA receptor antagonist, acting at the polyamine site

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-56-2
Molecular Formula C21H27NO2
Molecular Weight (g/mol) 325.45 g/mol
MDL Number MFCD01529999
InChI Key UYNVMODNBIQBMV-UHFFFAOYSA-N
Synonym ifenprodil, vadilex, ifenprodil tartrate, dilvax, creocral, ifenprodil inn:dcf, ifenprodilum inn-latin, unii-r8oe3p6o5s, 4-2-4-benzylpiperidin-1-yl-1-hydroxypropyl phenol, r8oe3p6o5s
PubChem CID 3689
IUPAC Name 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
SMILES CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3

Specifications

Color White
Quantity 50 mg
Merck Index 14,4895
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ifenprodil hemitartrate
EINECSNumber 245-491-4
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.