Learn More
Specifications
Specifications
| Concentration | 0.2 mg/ml |
| For Use With (Application) | Antibody Competition |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| Gene ID (Entrez) | 3428 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | ifi-16, IFNGIP1gamma-interferon-inducible protein 16, interferon, gamma-inducible protein 16, interferon-gamma induced protein IFI 16, Interferon-inducible myeloid differentiation transcriptional activator, MGC9466, PYHIN2 |
| Gene Symbol | IFI16 |
| Research Category | Cytokine Research |
| Protein | IFI16 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
