Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFI16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551690 Supplier Invitrogen™ Supplier No. PA551690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEKFTPKKII AIANYVCRNG FLEVYPFTLV ADVNADRNME IPKGLIRSAS VTPKINQLCS QTKGSFVNGV FEVHKKNVRG EFTYYEIQDN TGKMEVVVHG RLTTINCEEG DKLKLTCFEL APKSGNTGEL RSVIHSHIKV IK Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 39%.

IFI16 (gamma-inteferon-inducible protein 16) binds double-stranded DNA. It binds preferentially to supercoiled DNA and cruciform DNA structures. It seems to be involved in transcriptional regulation and as a transcriptional repressor. IFI16 may have a role in the regulation of hematopoietic differentiation through activation of unknown target genes. It controls cellular proliferation by modulating the functions of cell cycle regulatory factors including p53/TP53 and the retinoblastoma protein. IFI16 is also involved in innate immune response by recognizing viral dsDNA in the cytosol and nucleus. After binding to viral DNA in the cytoplasm, IFI16 recruits TMEM173/STING and mediates the induction of IFN-beta. It is necessary to activate the IFR3 signaling cascade suring human herpes simplex virus 1 infection and promotes the assembly of heterochromatin on herpesviral DNA and inhibiton of viral immediate-early gene expression and replication. IFI16 is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFI16
Gene Accession No. Q16666
Gene Alias gamma-interferon-inducible protein 16; IFI 16C; IFI16; Ifi-16; IFNGIP1; interferon gamma inducible protein 16; interferon, gamma-inducible protein 16; interferon-gamma induced protein IFI 16; interferon-inducible myeloid differentiation transcriptional activator; PYHIN2
Gene Symbols IFI16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFI16. Recombinant protein control fragment (Product #RP-101035).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3428
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.