Learn More
Invitrogen™ Ifi202 Polyclonal Antibody

Description
Ifi202 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Ifi202 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Ifi202b |
| Gene Accession No. | Q9R002 |
| Gene Alias | Ifbip-1; Ifi202; Ifi-202; Ifi202a; Ifi202b; interferon activated gene 202A; interferon activated gene 202B; interferon-activable protein 202; Interferon-activatable protein 202a (Ifi-202a) (Interferon-inducible protein p202a); interferon-inducible protein p202; Lupus susceptibility protein p202; p202 |
| Gene Symbols | Ifi202b, LOC100044068 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.