Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ifi204 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $426.00
Specifications
Antigen | Ifi204 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227153SS
![]() |
Novus Biologicals
NBP227153SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227153
![]() |
Novus Biologicals
NBP227153 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Ifi204 Polyclonal specifically detects Ifi204 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ifi204 | |
Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
Ifi16, Ifi204 interferon activated gene 204, p204 | |
IFI204 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in mouse. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse | |
P0DOV2 | |
15951 | |
A portion of amino acids 70-120 of mouse Ifi204 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title