Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI27 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3871 Supplier Bioss Supplier No. BS15549R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Promotes cell death. Mediates IFN-induced apoptosis characterized by a rapid and robust release of cytochrome C from the mitochondria and activation of BAX and caspases 2, 3, 6, 8 and 9.

Specifications

Antigen IFI27
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IFI27
Gene Accession No. P40305
Gene Alias 2310061N23Rik; FAM14D; IFI27; interferon alpha inducible protein 27; Interferon alpha-induced 11.5 kDa protein; Interferon alpha-inducible protein 27, mitochondrial; interferon, alpha-inducible protein 27; Interferon-stimulated gene 12a protein; ISG12; ISG12(a); ISG12A; p27
Gene Symbols IFI27
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IFI27.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.