Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFI30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109845 Supplier Invitrogen™ Supplier No. PA5109845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTLSPLLLFL PPLLLLLDVP TAAVQASPLQ ALDFFGNGPP VNYKTGNLYL RGPLKKSNAP LVNVTLYYEA LCGGCRAFLI RELFPTWLLV MEILNVTLVP YGNAQEQNVS GRWEFKCQHG EEECKFNKVE ACVLDELDME LAFLTIVCME EFEDMERSLP LCLQLYAPGL SPDTIMECAM GDRGMQLMHA NAQRTDALQP PHEYVPWVTV NGKPLEDQTQ LLTLVCQLYQ GKKPDVCPSS TSSLRSVCFK.

The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI30
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IFI30
Gene Accession No. P13284
Gene Alias Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain
Gene Symbols IFI30
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human IFI30 (NP_006323.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10437
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.