Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFI44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597852
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597852 Supplier Invitrogen™ Supplier No. PA597852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFI44 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

IFI-44 (interferon-induced protein 44), also known as p44 or MTAP44 (microtubule-associated protein 44), is a 444 amino acid protein that localizes to the cytoplasm and, upon induction by IFN-βs, aggregates to form microtubular structures. Human IFI-44 shares 97% sequence similarity with its chimp counterpart, suggesting a conserved role between species. The gene encoding IFI-44 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI44
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IFI44
Gene Accession No. Q8BV66, Q8TCB0
Gene Alias A430056A10Rik; AW261460; Ifi44; interferon induced protein 44; interferon-induced protein 44; interferon-induced, hepatitis C-associated microtubular aggregate protein (44kD); microtubule-associated protein 44; Mtap44; p44; TBC/LysM-associated domain containing 5; TLDC5
Gene Symbols IFI44
Host Species Rabbit
Immunogen Recombinant Human Interferon-induced protein 44 protein (101-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10561, 99899
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.