Learn More
Invitrogen™ IFI44L Polyclonal Antibody

Description
Immunogen sequence: IIDEQLVCRL SKTDIFIICR DNKIYLDKMI TRNLKLRFYG HRQYLECEVF RVEGIKDNLD DIKRIIKARE HRNRLL Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 33%.
Specifications
Specifications
| Antigen | IFI44L |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | IFI44L |
| Gene Accession No. | Q53G44 |
| Gene Alias | C1orf29; GS3686; H28; H-28; H28-1; H28c minor histocompatibility antigen 1; histocompatibility 28; HLA-HA28; IFI44L; IFL8; interferon induced protein 44 like; interferon-induced protein 44 like; interferon-induced protein 44-like; Minor histocompatibility antigen HA-28; NS1178 |
| Gene Symbols | IFI44L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.