Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFI44L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597853
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597853 Supplier Invitrogen™ Supplier No. PA597853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFI44L Polyclonal Antibody for Western Blot, ICC/IF, ELISA

The function remains known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI44L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IFI44L
Gene Accession No. Q53G44
Gene Alias C1orf29; GS3686; H28; H-28; H28-1; H28c minor histocompatibility antigen 1; histocompatibility 28; HLA-HA28; IFI44L; IFL8; interferon induced protein 44 like; interferon-induced protein 44 like; interferon-induced protein 44-like; Minor histocompatibility antigen HA-28; NS1178
Gene Symbols IFI44L
Host Species Rabbit
Immunogen Recombinant Human Interferon-induced protein 44-like protein (206-307AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10964
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.