Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFI44L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542829
Description
Peptide sequence: MEVTTRLTWN DENHLRKLLG NVSLSLLYKS SVHGGSIEDM VERCSRQGCT Sequence homology: Human: 100%.
The function remains known.
Specifications
| IFI44L | |
| Polyclonal | |
| Unconjugated | |
| IFI44L | |
| C1orf29; GS3686; H28; H-28; H28-1; H28c minor histocompatibility antigen 1; histocompatibility 28; HLA-HA28; IFI44L; IFL8; interferon induced protein 44 like; interferon-induced protein 44 like; interferon-induced protein 44-like; Minor histocompatibility antigen HA-28; NS1178 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10964 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q53G44 | |
| IFI44L | |
| Synthetic peptide directed towards the N-terminal of human IFI44L (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction